Silicon ≥99.9985% (metals basis), powder, crystalline amorphous ∼1-20 µm

Supplier: Thermo Scientific
Warning

38715.30EA 372 EUR
38715.30
Silicon ≥99.9985% (metals basis), powder, crystalline amorphous ∼1-20 µm
Silicon
Formula: Si
MW: 28,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
UN: 1346
ADR: 4.1,III
Merck Index: 12,08635

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR