n-Pentane ≥99%, Spectrophotometric Grade, Riedel-de Haën™
Supplier: Honeywell Chemicals
|
Danger
|
Synonyms:
Pentane
154954-2LEA
216.34
EUR
154954-2L
n-Pentane ≥99%, Spectrophotometric Grade, Riedel-de Haën™
n-Pentane
- Spectrophotometric grade solvents are high purity solvents made specifically for applications using spectrophotometers.
Beilstein Number: 969132
RTECS Number: RZ9450000
UNSPSC Code: 12191502
Formula:
H₃C(CH₂)₃CH₃ MW: 72,15 g/mol Boiling Pt: 36-37 °C Melting Pt: -131 °C Density: 0,631 g/cm³ (15 °C) Flash Pt: <-35 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009498 CAS Number: 109-66-0 EINECS: 203-692-4 UN: 1265 ADR: 3,II Merck Index: 12,07255 |
Specification Test Results
Identity (IR) | complying |
Assay (GC) | Min. 99.0 % |
Refractive index (n 20/D) | 1.356 - 1.360 |
Non-volatile matter | Max. 0.0003 % |
Water (Karl Fischer) | Max. 0.01 % |
Absorbance at 400 nm | Max. 0.01 |
Absorbance at 245 nm | Max. 0.01 |
Absorbance at 220 nm | Max. 0.06 |
Absorbance at 210 nm | Max. 0.30 |
Absorbance at 200 nm | Max. 0.60 |
Absorbance at 195 nm | Max. 1.00 |
Vapor Pressure | 26.98 psi ( 55 °C) |
Vapor Pressure | 8.4 psi ( 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...