Sodium orthovanadate, activated

Supplier: BioVision

BIOV9471-5EA 130 EUR
BIOV9471-5
Sodium orthovanadate, activated
Sodium orthovanadate

Activated sodium orthovanadate is a depolymerised vanadate prepared from sodium orthovanadate that displays enhanced ability to inhibit endogenous phosphatases in a competitive manner. Preserves the phosphorylation state in cells and cell lysates. Completely reversible with EDTA or by dilution.


  • Protein tyrosine phosphatase (PTP) inhibitor
  • To be protected from air and moisture


Appearance: Liquid


Formulation: 100 mM solution in sterile distilled water with pH adjusted to 10


Caution: For research use only.

Formula: Na₃O₄V
MW: 183,91 g/mol
MDL Number: MFCD00003511
CAS Number: 13721-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR