Petroleum spirit, 140 - 180 °C, EMPLURA®, Supelco®

Supplier: Merck
Danger

Synonyms: Petroleum etherGasolinePetrol

M-Clarity™ quality level = MQ200

8.14563.1000EA 61.7 EUR
8.14563.9156 8.14563.1000
Petroleum spirit, 140 - 180 °C, EMPLURA®, Supelco®
Petroleum spirit
Boiling Pt: 140…180 °C (1013 hPa)
Melting Pt: < –15 °C
Density: 0,77 g/cm³ (20 °C)
Flash Pt: 24 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-82-1
EINECS: 265-151-9
UN: 1268
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR