Petroleum spirit, 40 - 80 °C, EMPLURA®, Supelco®

Supplier: Merck
Danger

Synonyms: Petroleum etherGasolinePetrol

M-Clarity™ quality level = MQ200

1.01773.6025EA 908 EUR
1.01773.6025 1.01773.1000
Petroleum spirit, 40 - 80 °C, EMPLURA®, Supelco®
Petroleum spirit
Boiling Pt: 40…80 °C (1013 hPa)
Density: 0,665 g/cm³ (15 °C)
Flash Pt: <–30 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR