Petroleum spirit, 50 - 70 °C, EMPLURA®, Supelco®

Supplier: Merck
Danger

Synonyms: Petroleum etherGasolinePetrol

M-Clarity™ quality level = MQ200

1.00910.1000EA 85.3 EUR
1.00910.1000 1.00910.6025
Petroleum spirit, 50 - 70 °C, EMPLURA®, Supelco®
Petroleum spirit
Boiling Pt: 50…70 °C (1013 hPa)
Melting Pt: < –20 °C
Density: 0,654…0.67 g/cm³ (20 °C)
Flash Pt: <–21 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR