Starch, hydrolysed acc. Smithies

Supplier: Molekula

MOLEM55960455EA 0 EUR
MOLEM55960455
Starch, hydrolysed acc. Smithies
Starch
Formula: (C₆H₁₀O₅)n
MW: 162,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-25-8
EINECS: 232-679-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR