Polyethylene glycol 300, TECHNICAL
Supplier: VWR Chemicals
Synonyms:
PEG 300
26600.293EA
50.4
EUR
26600.293
Polyethylene glycol 300, TECHNICAL
Polyethylene glycol 300
Formula:
H(OCH₂CH₂)nOH MW: 300 g/mol Melting Pt: –15…–10 °C Density: 1,13 g/cm³ (20 °C) Flash Pt: 220 °C (open cup) Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 EINECS: 500-038-2 |
Specification Test Results
Identification | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...