Calcium-D-gluconate monohydrate

Synonyms: Calcium gluconate monohydrateGluconic acid calcium salt monohydrateD-Gluconic acid calcium salt monohydrate

EDQMC0300000EA 221 EUR
EDQMC0300000
Calcium-D-gluconate monohydrate
Calcium D-gluconate monohydrate
Formula: C₁₂H₂₂CaO₁₄·H₂O
MW: 448,39 g/mol
Melting Pt: 178 °C
Storage Temperature: Ambient
MDL Number: MFCD00150787
CAS Number: 66905-23-5
EINECS: 206-075-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR