(±)-Phenylsuccinic acid
Supplier: Molekula
Synonyms:
Phenylsuccinic acid
, (±)-Phenyl-1,4-butanedioic acid
MOLE21959847-100GEA
97.1
EUR
MOLE21959847-100G
(±)-Phenylsuccinic acid
(±)-Phenylsuccinic acid
Formula:
C₁₀H₁₀O₄ MW: 194,19 g/mol Boiling Pt: 368 °C (1013 hPa) Melting Pt: 167…169 °C |
MDL Number:
MFCD00004256 CAS Number: 635-51-8 EINECS: 211-238-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...