(±)-1-Hexen-3-ol ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(±)-Hex-1-en-3-ol
, hex-1-En-3-ol
A19672.18EA
142
EUR
A19672.18
A19672.09
(±)-1-Hexen-3-ol ≥98%
(±)-1-Hexen-3-ol
Formula:
C₆H₁₂O MW: 100,16 g/mol Boiling Pt: 132…134 °C Melting Pt: –72 °C Density: 0,837 Flash Pt: 35 °C (95 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004581 CAS Number: 4798-44-1 EINECS: 225-355-0 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...