Guanidinium hydrochloride 8 mol/l in aqueous solution for biotechnology
|
Warning
|
Synonyms:
Guanidinium chloride
, Guanidine hydrochloride
, Guanidine chloride
, Guanadine hydrochloride
J842-4LEA
0
EUR
J842-4L
Guanidinium hydrochloride 8 mol/l in aqueous solution for biotechnology
Guanidinium hydrochloride
A convenient premixed solution of the biotechnology grade powder easily diluted for almost any RNA or protein purification procedure.
Formula:
NH₂C(=NH)NH₂·HCl MW: 95,53 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013026 CAS Number: 50-01-1 EINECS: 200-002-3 |
Specification Test Results
Absorbance @ 260 nm | <0.05 |
Absorbance @ 280 nm | <0.15 |
Conductivity @25°C (1:100, Water) | 8 - 10 mmhos |
Specific Gravity | 1.1849 - 1.1949 g/ml |
Titration | 7.5 - 8.5 Molar |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...