Acrylamide 40% in aqueous solution, 4x crystallized

Danger

Synonyms: Prop-2-enamide

A0962.1000EA 0 EUR
A0962.1000
Acrylamide 40% in aqueous solution, 4x crystallized
Acrylamide (monomer)
Formula: C₃H₅NO
MW: 71,08 g/mol
MDL Number: MFCD00008032
CAS Number: 79-06-1
EINECS: 201-173-7
UN: 3426
ADR: 6.1,III
https://ie.vwr.com/stibo/web/std.lang.all/13/21/2301321.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR