Hydrochloric acid 32%
Supplier: Honeywell Chemicals
|
Danger
|
40233-2.5LEA
136.19
EUR
40233-2.5L
Hydrochloric acid 32%
Hydrochloric acid
Formula:
HCl MW: 36,46 g/mol Boiling Pt: 110 °C (1013 hPa) Melting Pt: –30 °C Density: 1,18 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...