5(S),12(R)-Dihydroxyeicosa-6E,8E,10E,14Z-tetraenoic acid 50 µg/ml in ethanol , 99% (by HPLC)
ALEXBMLDTR512-0050EA
329
EUR
ALEXBMLDTR512-0050
ALEXBMLDTR512-1000
5(S),12(R)-Dihydroxyeicosa-6E,8E,10E,14Z-tetraenoic acid 50 µg/ml in ethanol , 99% (by HPLC)
5(S),12(R)-Dihydroxyeicosa-6E,8E,10E,14Z-tetraenoic acid
Biosynthesis via non enzymatic and enzymatic hydrolysis of LTA₄ and by myeloperoxidase-dependent metabolism of cysteinyl leukotrienes. Neutrophil chemotaxin in the guinea pig dermis. Metabolite of LTB₄ in rat kidney produced enzymatically by a novel isomerase reaction.
Formula:
C₂₀H₃₂O₄ |
CAS Number:
71652-82-9 |
Specification Test Results
Purity | ≥99% (HPLC) |
Concentration | 50 µg/ml |
Formulation | Oil dissolved in ethanol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...