9-Methyl-3-((2-methyl-1H-imidazol-1-yl)methyl)-2,3-dihydro-1H-carbazol-4(9H)-one ≥98% (by NMR), white solid
Supplier: Enzo Life Sciences
|
Danger
|
Synonyms:
Ondansetron
ALEXBML-NS145-0050EA
513
EUR
ALEXBML-NS145-0050
9-Methyl-3-((2-methyl-1H-imidazol-1-yl)methyl)-2,3-dihydro-1H-carbazol-4(9H)-one ≥98% (by NMR), white solid
9-Methyl-3-((2-methyl-1H-imidazol-1-yl)methyl)-2,3-dihydro-1H-carbazol-4(9H)-one
Serotonin receptor ligand
A serotonin 5-HT3 receptor antagonist. Clinically effective anti-emetic especially in chemotherapy-induced nausea.
Soluble in DMSO (10 mg/ml warm).
Formula:
C₁₈H₁₉N₃O MW: 293,36 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00833882 CAS Number: 99614-02-5 |
Specification Test Results
Purity | ≥98% (NMR) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...