Calcium pantothenate ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: D-Calcium pantothenateCalcium D-PantothenateD-Pantothenic acid calcium saltCalcium D-(+)-pantothenateCalcium (R)-3-(2,4-dihydroxy-3,3-dimethylbutanamido)propanoateVitamin B5 calcium salt

ALEXALX460003G010EA 60.8 EUR
ALEXALX460003G010
Calcium pantothenate ≥98% (by HPLC)
Calcium pantothenate

Precursor for coenzyme A biosynthesis.


Soluble in water. Also soluble in glycerol; slightly soluble in 100% ethanol or methanol; insoluble in acetone or ether.

Formula: C₁₈H₃₂CaN₂O₁₀
MW: 476,54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002766
CAS Number: 137-08-6
EINECS: 205-278-9
https://ie.vwr.com/stibo/web/std.lang.all/07/47/45390747.jpg

Order Now

Specification Test Results

Purity (HPLC) ≥98%
Identity Determined by IR
Appearance White to off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR