Adenosine 5'-monophosphate disodium salt (AMP disodium salt) ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 5′-AMP-Na2

ALEXALX480014G005EA 103 EUR
ALEXALX480014G005
Adenosine 5'-monophosphate disodium salt (AMP disodium salt) ≥98% (by HPLC)
Adenosine 5'-monophosphate disodium salt (AMP disodium salt)

Soluble in water or aqueous buffers.

Formula: C₁₀H₁₂N₅Na₂O₇P
MW: 391,2 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00065023
CAS Number: 4578-31-8
EINECS: 224-961-2
https://ie.vwr.com/stibo/web/std.lang.all/07/61/45390761.jpg

Order Now

Specification Test Results

Purity (HPLC) ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR