3,5-Dimethyl-1-adamantanamine hydrochloride ≥98% (by argentometric titration)

Supplier: Enzo Life Sciences

Synonyms: 1-Amino-3,5-dimethyladamantane HydrochlorideMemantine Hydrochloride

ALEXALX550363M050EA 126 EUR
ALEXALX550363M050
3,5-Dimethyl-1-adamantanamine hydrochloride ≥98% (by argentometric titration)
3,5-Dimethyl-1-adamantanamine hydrochloride

NMDA receptor antagonist.


Low-affinity NMDA receptor antagonist. Stimulates dopamine release.

Formula: C₁₂H₂₁N
MW: 215,77 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00214336
CAS Number: 41100-52-1
EINECS: 255-219-6
https://ie.vwr.com/stibo/web/std.lang.all/08/52/45390852.jpg

Order Now

Specification Test Results

Purity (argentmetric titration) ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR