D-(-)-Mannitol, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, FCC, E421, SAFC®
Synonyms:
Mannitol
, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
1.05980.2500EA
0
EUR
1.05980.2500
1.05980.9050
D-(-)-Mannitol, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, FCC, E421, SAFC®
D(-)-Mannitol
Formula:
HOH₂C(CH(OH))₄CH₂OH MW: 182,17 g/mol Boiling Pt: 290…295 °C (4 hPa) Melting Pt: 164…169 °C Density: 1,49 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064287 CAS Number: 69-65-8 EINECS: 200-711-8 Merck Index: 13,05769 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...