D-(-)-Mannitol, fine powder, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, FCC, E421, SAFC®

Supplier: MERCK PRODUCTION CHEMICALS

Synonyms: Mannitol(2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol

1.05988.9050EA 0 EUR
1.05988.9050
D-(-)-Mannitol, fine powder, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, FCC, E421, SAFC®
D(-)-Mannitol
Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182,17 g/mol
Boiling Pt: 290…295 °C (4 hPa)
Melting Pt: 164…169 °C
Density: 1,49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064287
CAS Number: 69-65-8
EINECS: 200-711-8
Merck Index: 13,05769
https://ie.vwr.com/stibo/web/std.lang.all/06/13/16640613.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR