L(+)-Valine

Supplier: EDQM

Synonyms: (S)-(+)-ValineValineH-Val-OH

EDQMV0030000EA 200 EUR
EDQMV0030000
L(+)-Valine
L(+)-Valine
Formula: C₅H₁₁NO₂
MW: 117,15 g/mol
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 86 °C
Density: 1,284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064220
CAS Number: 72-18-4
EINECS: 200-773-6
Merck Index: 12,10046

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR