L(+)-Histidine 98.5-101.5%, high purity
Synonyms:
L-3-Imidazol-4-ylalanine
, (S)-(+)-Histidine
, (S)-2-Amino-3-(4-imidazolyl)propionic acid
, NSC 137773
, L-Histidine
, H-His-OH
1B1164-100GEA
83.8
EUR
1B1164-100G
1B1164-1KG
L(+)-Histidine 98.5-101.5%, high purity
L(+)-Histidine
Formula:
C₆H₉N₃O₂ MW: 155,16 g/mol Melting Pt: 272…273 °C Density: 1,44 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064315 CAS Number: 71-00-1 EINECS: 200-745-3 Merck Index: 13,04741 |
Specification Test Results
Arsenic |
|
Chloride |
|
Heavy Metals |
|
Iron |
|
Loss on Drying |
|
Purity (Dry Basis) | 98.5 – 101.5% |
Residue after Ignition |
|
Specific Rotation | 12.4 – 14° |
Sulphate |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...