L(-)-Tryptophan
Synonyms:
L-α-Amino-3-indolepropionic acid
, (S)-2-Amino-3-(3-indolyl)propionic acid
, H-Trp-OH
, L-Tryptophan
, Tryptophan
EDQMT2610000EA
231
EUR
EDQMT2610000
L(-)-Tryptophan
L(-)-Tryptophan
Formula:
C₁₁H₁₂N₂O₂ MW: 204,23 g/mol Melting Pt: 290 °C Density: 1,34 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064340 CAS Number: 73-22-3 EINECS: 200-795-6 Merck Index: 12,09929 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...