SB-431542 5 mM in DMSO ≥98% (by HPLC), EZSolution™, sterile filtered

Supplier: BioVision

Synonyms: 4-[4-(1,3-benzodioxol-5-yl)-5-pyridin-2-yl-1H-imidazol-2-yl]benzamide

BIOV1674-1EA 124 EUR
BIOV1674-1 BIOV1674-5
SB-431542 5 mM in DMSO ≥98% (by HPLC), EZSolution™, sterile filtered
SB-431542

A potent ALK5 inhibitor.


Caution: For Research Use Only! Not For Use in Humans

Formula: C₂₂H₁₈N₄O₃
Storage Temperature: Freezer
MDL Number: MFCD11045982
CAS Number: 301836-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR