Trifluoromethanesulphonic anhydride

Supplier: Apollo Scientific
Danger

Synonyms: Triflic anhydride

APOSPC7420-250GEA 109 EUR
APOSPC7420-250G
Trifluoromethanesulphonic anhydride
Trifluoromethanesulphonic anhydride
Formula: C₂F₆O₅S₂
MW: 282,14 g/mol
Boiling Pt: 83 °C (1013 hPa)
Density: 1,67 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000408
CAS Number: 358-23-6
EINECS: 206-616-8
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR