Isoamyl alcohol ≥98.5% ACS, Riedel-de Haën™

Warning

Synonyms: 3-Methyl-1-butanolIsopentyl alcohol3-Methylbutanoliso-Pentanol

320021-1LEA 161.57 EUR
320021-1L
Isoamyl alcohol ≥98.5% ACS, Riedel-de Haën™
Isoamyl alcohol

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Formula: (CH₃)₂CHCH₂CH₂OH
MW: 88,15 g/mol
Boiling Pt: 131 - 132 °C
Melting Pt: -117 °C
Density: 0,81 g/cm³ (20 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00002934
CAS Number: 123-51-3
EINECS: 204-633-5
UN: 1105
ADR: 3,III
Merck Index: 13,05214
https://ie.vwr.com/stibo/web/std.lang.all/15/69/2301569.jpg

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 98.50 %
Non-volatile matter Max. 0.003 %
Water (Karl Fischer) Max. 0.5 %
Titrable Acid Max. 0.002 meq/g
Acids, esters (as amyl acetate) Max. 0.2 %
Carbonyl Compounds (as HCHO) Max. 0.1 %
Vapor Density 3 (vs air)
Refractive Index n20/D 1.406 (lit.)
Explosion Limit 1.2-9 %, 100 °F

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR