cis-1,4-Dichloro-2-butene 95%

Supplier: Thermo Scientific
Danger

A18956.22EA 256 EUR
A18956.22
cis-1,4-Dichloro-2-butene 95%
cis-1,4-Dichloro-2-butene

Purity: 95%
CAS No.: 1476-11-5
Molecular Formula: C4H6Cl2
Formula Weight: 125.00
Storage Temperature: 2°C to 8°C
Flash Point: 55°(132°F)
Refractive Index: 1.4900
Melting Point: -48°
Boiling Point: 151-153°
Beilstein Reference: 1719692
UN No.: 3390
MDL No.: MFCD00062950

Formula: C₄H₆Cl₂
MW: 125 g/mol
Boiling Pt: 151…153 °C
Melting Pt: –48 °C
Density: 1,188
Flash Pt: 55 °C (132 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00062950
CAS Number: 1476-11-5
EINECS: 216-021-5
UN: 3390
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR