Acrylamide ≥99.9%, Proteomics Grade
Supplier: VWR Chemicals
|
Danger
|
Synonyms:
Prop-2-enamide
M120-500GEA
0
EUR
M120-500G
Acrylamide ≥99.9%, Proteomics Grade
Acrylamide (monomer)
VWR offers an extensive line of acrylamide and bis-acrylamide pre-weighed powder blends, pre-mixed stock solutions, and ready to use solutions for customised polyacrylamide gel electrophoresis of proteins and PAGE of nucleic acids. The VWR acrylamide and bis-acrylamide powders provide the flexibility to prepare solutions having concentrations and ratios for all electrophoresis applications. Liquid blends minimise the handling of neurotoxic acrylamide.
- Ultra-pure powders with acrylamide purity > 99.9%
- Convenient, ready to use
- Eliminates hazardous powdered acrylamide handling
Formula:
C₃H₅NO MW: 71,08 g/mol Boiling Pt: 232 °C (1013 hPa) Melting Pt: 84,5 °C Density: 1,122 g/cm³ (20 °C) Flash Pt: 138 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008032 CAS Number: 79-06-1 EINECS: 201-173-7 UN: 2074 ADR: 6.1,III |
Specification Test Results
Conductivity (40%, Water) |
|
DNase | none detected |
Electrophoresis | Pass |
Free Acrylic Acid |
|
Appearance | White free-flowing, uniform crystals |
Insolubles |
|
Iron |
|
Lead |
|
Magnesium |
|
Melting Point | 84 – 86 °C |
Moisture (KF) |
|
pH (10%, 0.1 M NaCl) @25 °C |
|
Protease | none detected |
Purity | ≥99.9% |
RNase | none detected |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...