3-Chloro-2,4,5,6-tetrafluorobenzoyl chloride ≥98%
Supplier: Thermo Scientific
|
Danger
|
L20166.06EA
83.9
EUR
L20166.06
L20166.14
L20166.03
3-Chloro-2,4,5,6-tetrafluorobenzoyl chloride ≥98%
3-Chloro-2,4,5,6-tetrafluorobenzoyl chloride
Formula:
C₇Cl₂F₄O MW: 246,98 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03001175 CAS Number: 292621-58-0 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...