3-(Trifluoromethoxy)thiophenol

Supplier: Apollo Scientific

APOSPC9232-1GEA 73.9 EUR
APOSPC9232-1G
3-(Trifluoromethoxy)thiophenol
3-(Trifluoromethoxy)thiophenol
Formula: C₇H₅F₃OS
MDL Number: MFCD01320799
CAS Number: 220239-66-7
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR