Hydroquinone, Fotopur®
Supplier: Merck
|
Danger
|
Synonyms:
1,4-Dihydroxybenzene
, Quinol
1.04610.9026EA
0
EUR
1.04610.9026
Hydroquinone, Fotopur®
Hydroquinone
Formula:
C₆H₄(OH)₂ MW: 110,11 g/mol Boiling Pt: 285 °C (1013 hPa) Melting Pt: 172 °C Density: 1,332 g/cm³ (20 °C) Flash Pt: 165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002339 CAS Number: 123-31-9 EINECS: 204-617-8 UN: 3077 ADR: 9,III Merck Index: 13,04833 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...