3',5'-Dihydroxyacetophenone 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-(3,5-Dihydroxyphenyl)ethanone
, 1-(3,5-Dihydroxyphenyl)ethan-1-one
ACRO208630250EA
259
EUR
ACRO208630250
3',5'-Dihydroxyacetophenone 97%
3',5'-Dihydroxyacetophenone
Formula:
C₈H₈O₃ MW: 152,15 g/mol Boiling Pt: 296 °C (1013 hPa) Melting Pt: 143…147 °C |
MDL Number:
MFCD00002290 CAS Number: 51863-60-6 EINECS: 257-480-1 |
Specification Test Results
Appearance | Off-white to brown Powder |
Infrared spectrum | Conforms |
Melting point | 143 to 148 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...