4-Hexylresorcinol
|
Warning
|
Synonyms:
4-Hexylbenzene-1,3-diol
EDQMH0420000EA
231
EUR
EDQMH0420000
4-Hexylresorcinol
4-Hexylresorcinol
Formula:
C₁₂H₁₈O₂ MW: 194,27 g/mol Boiling Pt: 333 °C (1013 hPa) Melting Pt: 64…67 °C Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002284 CAS Number: 136-77-6 EINECS: 205-257-4 Merck Index: 12,04750 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...