Toluene ≥99.5% ACS, Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

Synonyms: PhenylmethaneMethylbenzene

320552-1LEA 50.07 EUR
320552-1L
Toluene ≥99.5% ACS, Riedel-de Haën™
Toluene
Formula: C₆H₅CH₃
MW: 92,14 g/mol
Boiling Pt: 110,6 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0,867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 12,09667
https://ie.vwr.com/stibo/web/std.lang.all/61/73/766173.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR