Benzyl butyl phthalate-[ring-D₄] (98% D)
Supplier: LGC Standards PROMOCHEM
Synonyms:
Benzyl butyl phthalate-[ring-²H₄]
PROOCILDLM13690.1EA
582
EUR
PROOCILDLM13690.1
CAMBDLM-1369-0.1
Benzyl butyl phthalate-[ring-D₄] (98% D)
Benzyl butyl phthalate-[ring-D4]
Formula:
C₁₉H₁₆D₄O₄ |
CAS Number:
93951-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...