Sodium sulphate
Synonyms:
Sulphuric acid disodium salt
, Bisodium sulphate
, Disodium sulphate
PFIZS/6640/53EA
0
EUR
PFIZS/6640/53
NOVSS/6640/53
FISHS/6640/53
XXXXS/6640/63
Sodium sulphate
Sodium sulphate
Formula:
Na₂SO₄ MW: 142,04 g/mol Boiling Pt: 1429 °C (1013 hPa) Melting Pt: 888 °C Density: 2,7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003504 CAS Number: 7757-82-6 EINECS: 231-820-9 Merck Index: 12,08829 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...