Dimethyl sulphoxide, Supelco®

Supplier: Merck

Synonyms: DMSOMethyl sulphoxide

M-Clarity™ quality level = MQ100

1.09678.0100EA 98.4 EUR
1.09678.0100
Dimethyl sulphoxide, Supelco®
Dimethyl sulphoxide
Formula: (CH₃)₂SO
MW: 78,14 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 18,5 °C
Density: 1,101 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://ie.vwr.com/stibo/web/std.lang.all/77/31/2297731.jpg

Order Now

Specification Test Results

Solubility 1000 g/l (20 °C)
Melting Point 18.4 °C
Molar Mass 78.1 g/mol
Boiling Point 189 °C (1013 hPa)
Vapor Pressure 0.6 hPa (20 °C)
Flash Point 87 °C
Refractive Index 1.48 (20 °C)
Explosion Limit 1.8 - 63.0 %(V)
Density 1.10 g/cm3 (20 °C)
Ignition Point 301 °C
Saturation concentration 8.0 g/m3 (20 °C)
Viscosity kinematic 2.14 mm2/s

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR