Dimethyl sulphoxide for synthesis, Supelco®
Synonyms:
DMSO
, Methyl sulphoxide
8.02912.9999
Dimethyl sulphoxide for synthesis, Supelco®
Dimethyl sulphoxide
Formula:
(CH₃)₂SO MW: 78,14 g/mol Boiling Pt: 189 °C (1013 hPa) Melting Pt: 18,5 °C Density: 1,101 g/cm³ (20 °C) Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002089 CAS Number: 67-68-5 EINECS: 200-664-3 Merck Index: 13,03285 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...