2-Chloropyrazine
Supplier: Molekula
MOLEM87212357EA
0
EUR
MOLEM87212357
2-Chloropyrazine
2-Chloropyrazine
Formula:
C₄H₃ClN₂ MW: 114,53 g/mol Boiling Pt: 153 °C (1013 hPa) Density: 1,28 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006124 CAS Number: 14508-49-7 EINECS: 238-517-0 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...