Bottles, narrow neck, allround
Supplier: Rixius
RIXI10501005018OSVEA 113 EUR
RIXI10501005018OSV
Bottles, narrow neck, allround
Bottles Glass Bottles
Amber glass without screw cap.
  • Good chemical and physical properties
  • For applications that do not involve high thermal stresses
  • Round thread in accordance with DIN 168
Excellent for sampling, storing and transporting liquids. Amber coloured to protect light-sensitive materials.