Laboratory freezer, Mediline, LGv 5010
Supplier: Liebherr
Mediline
471-1213EA 0 EUR
471-1213
Laboratory freezer, Mediline, LGv 5010
Freezers Laboratory / General Purpose Freezers
The Liebherr LGV 5010 is a large capacity laboratory grade freezer. The LGV 5010 comes from the Liebherr Mediline laboratory range. This large capacity freezer is purpose built for the laboratory market for operation with constant use in demanding working conditions. The freezer features a RS485 interface, no frost technology, capturing heat energy from the refrigeration cycle allowing a gentle automatic defrost cycle, preventing ice building up.
  • The advanced electronic controller comes with a variety of visual and audible fault warning alarms and can store three alarm events
  • Maximum temperature fluctuation: 4,9 °C
  • No frost technology, automatic defrost
  • Electronic controller (which can be set in the temperature range)
  • Advanced comfort digital electronic temperature display
  • Visual and audio fault warning alarms (high/low temp and door open)
  • Integrated data memory, stores three alarm events
  • Environmentally friendly refrigerant used, R 290
  • Comes on lockable castors
  • Power switch with safety feature, must be held down 5 seconds to prevent accidental power off