Ascorbate oxidase (from Cucurbita sp.)
Supplier: Thermo Fisher Scientific
J65782.4IEA 56.8 EUR
J65782.4I
Ascorbate oxidase (from Cucurbita sp.)
Enzymes
Ascorbate oxidase, from Cucurbita sp. can be used to study oxidative stress and heat stress response and tolerance.
It is also used in ascorbic acid assays to study the heat stress response of arabidopsis and useful for enzymatic determination of ascorbic acid and for eliminating the interference of ascorbic acid in clinical analysis.
Caution: Keep the container tightly closed. Store away from oxidizing agent.