D-(-)-Sorbitol 91.0-100.5% (dry basis), Ultrapure

Supplier: Thermo Scientific

Synonyms: SorbitolD(-)-Glucitol(Sorbitol)

J21710.A7EA 145 EUR
J21710.A7 J21710.36 J21710.A1
D-(-)-Sorbitol 91.0-100.5% (dry basis), Ultrapure
D(-)-Sorbitol
Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182,17 g/mol
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004708
CAS Number: 50-70-4
EINECS: 200-061-5
https://ie.vwr.com/stibo/web/std.lang.all/97/21/2299721.jpg

Order Now

Specification Test Results

Assay (Dry Basis) 91.0 to 100.5%
Assay Reducing Sugars ≤0.3%
Impurity Content Limit of Nickel ≤1 ppm
Impurity Content Total Molds and Yeasts ≤100 cfu/g
Water Content (Karl Fischer Titration) ≤1.5%
pH (10%, H₂O) 3.5 - 7.0
Identification (FTIR) Conforms
Identity (A, B) Passes Tests
Total Aerobic Plate Count ≤1000 cfu/g
Residue on Ignition ≤0.1%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR