Ethyl acetate ≥99%, Reagent Grade

Danger

Synonyms: Acetic acid ethyl ester

45780-25LEA 489.65 EUR
45780-25L
Ethyl acetate ≥99%, Reagent Grade
Ethyl acetate
Formula: CH₃COOC₂H₅
MW: 88,11 g/mol
Boiling Pt: 77,1 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0,902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
https://ie.vwr.com/stibo/web/std.lang.all/20/72/722072.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR