Diethyl ether ≥99.9% inhibitor-free, CHROMASOLV™ for HPLC, Riedel-de Haën™
|
Danger
|
Synonyms:
Ether
, 2,2'-Oxybisethane
, Ethoxyethane
, Ethyl Ether
, 2,2'-Oxydiethane
309966-1LEA
99.18
EUR
309966-1L
309966-100ML
Diethyl ether ≥99.9% inhibitor-free, CHROMASOLV™ for HPLC, Riedel-de Haën™
Diethyl ether
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Formula:
(CH₃CH₂)₂O MW: 74,12 g/mol Boiling Pt: 35 °C Melting Pt: -117,4 °C Density: 0,713 g/cm3 Flash Pt: -45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011646 CAS Number: 60-29-7 EINECS: 200-467-2 UN: 1155 ADR: 3,I REACH: 01-2119535785-29 |
![]() |
Specification Test Results
Identity (IR) | complying |
Assay (GC) | min. 99.90 % |
Non-volatile matter | max. 0.0003 % |
Water (Karl Fischer) | max. 0.03 % |
Peroxides (as H₂O₂) | max. 1 ppm |
Absorbance at 400 nm | max. 0.01 |
Absorbance at 300 nm | max. 0.01 |
Absorbance at 275 nm | max. 0.04 |
Absorbance at 250 nm | max. 0.20 |
Absorbance at 218 nm | max. 1.00 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...