Genesis tubing sealers
PAWB428-SE340EA 4300 EUR
PAWB428-SE340 PAWB428-SE640
Genesis tubing sealers
Heat Sealers Tube Sealers
Genesis tubing sealers provide rapid and highly integral tubing seals using lightweight and portable equipment.
  • Highly integral seals
  • Easy to disconnect
  • Seals dry or liquid filled tubing
  • Widely used in blood banking applications
  • Battery or AC powered options available
When the trigger lever is pulled, the Genesis device initiates strong high-frequency energy which instantly melts the thermoplastic Tygon® tubing
The seal is designed for easy separation. After the sealing cycle has completed, the tubing can be aseptically parted by pulling on tube itself. No other tools are required and sterility of each side is completely maintained.
Depending on your requirements, you can select an AC powered or a battery powered system. The AC powered Model SE340 is equipped with two sealing heads. One is a fixed head mounted on the base, while the other is a tethered hand-held unit. The battery powered Model SE640 comes with a tethered head only, it does not have a fixed head.
The battery powered unit provides portability in areas without AC power.