Boiling flasks, flat bottom, with short neck, KIMAX®
SCERSP25055-300EA 1120 EUR
SCERSP25055-300 SCERSP25055-125 SCERSP25055-500 SCERSP25055-1000 SCERSP25055-250
Boiling flasks, flat bottom, with short neck, KIMAX®
Flasks Flat Bottom Flasks
KIMAX® flat bottom short neck boiling flasks with ST 24/40 joint, made of 33 expansion low extractable borosilicate glass.
  • Freestanding
  • Available in different capacities
Designed from ASTM Specification E1403, Type I, Class IV requirements.