Cyclohexane ≥99.8%, Laboratory Reagent

Supplier: Honeywell Chemicals
Danger

Synonyms: Hexamethylene

C100307-25LEA 601.61 EUR
C100307-25L C100307-5L C100307-1L C100307-2.5L C100307-150KG C100307-4X5L C100307-4X2.5L C100307-10L
Cyclohexane ≥99.8%, Laboratory Reagent
Cyclohexane

  • ≥95% for general laboratory use.

Formula: C₆H₁₂
MW: 84,16 g/mol
Boiling Pt: 81 °C
Melting Pt: 6,4 °C
Density: 0,78 g/cm3
Flash Pt: -18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
https://ie.vwr.com/stibo/web/std.lang.all/23/06/772306.jpg

Order Now

Specification Test Results

Appearance (form) complying
Appearance (color) complying
Purity (GC) min. 99.80 Fl.%
Non-volatile matter max. 0.005 %
IR spectrum complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR