Sodium sulphate, anhydrous ≥99.0%, powder ACS, Fluka™

Supplier: Honeywell Chemicals

Synonyms: Sulphuric acid disodium saltBisodium sulphateDisodium sulphate

238597-2.5KGEA 210.58 EUR
238597-2.5KG 238597-500G 238597-1KG
Sodium sulphate, anhydrous ≥99.0%, powder ACS, Fluka™
Sodium sulphate
Formula: Na₂SO₄
MW: 142,04 g/mol
Melting Pt: 884 °C
Density: 2,68 g/cm3
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
https://ie.vwr.com/stibo/web/std.lang.all/06/41/16640641.jpg

Order Now

Specification Test Results

Identity complying
Assay min. 99.0 %
pH (5 %, 25 °C) 5.2 - 9.2
Loss on ignition (800°C) max. 0.5 %
Water insoluble matter max. 0.01 %
Calcium (Ca) max. 0.01 %
Iron (Fe) max. 0.001 %
Potassium (K) max. 0.01 %
Magnesium (Mg) max. 0.005 %
Heavy metals (as Pb) max. 5 ppm
Chloride (Cl) max. 0.001 %
Phosphate (PO₄) max. 0.001 %
Total N max. 5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR